Reactant in the heterogeneously catalyzed etherification of glycerol, also in fritsch-wiechell rearrangement of 1-chlorovinyl sulfoxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Reaktionsmittel in der heterogen katalysierten Veretherifikation von Glycerin, auch in der Fritsch-Wchell-Neuanordnung von 1-Chlorovinylsulfoxiden.
Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von starken Oxidationsmitteln fernhalten.
RUO – Research Use Only
Allgemeine Referenzen:
- Yanlong Gu,; Ahmed Azzouzi,; Yannick Pouilloux,; François Jérôme,; Joël Barraulta. Heterogeneously catalyzed etherification of glycerol: new pathways for transformation of glycerol to more valuable chemicals. Green Chem. 2008, 10 164-167.
- Tsuyoshi Satoh,; Yasumasa Hayashi,; Koji Yamakawa. Fritsch-Wiechell Rearrangement of 1-Chlorovinyl Sulfoxides: A New Method for Synthesizing Acetylenes from Aldehydes with One-Carbon Homologation. Bulletin of the Chemical Society of Japan. 1993, 66 (6), 1866-1869.