Thermo Scientific Chemicals

Hexamethylacetone, 98%, Thermo Scientific Chemicals

Catalog number: H53464.14
25 g, Each
Thermo Scientific Chemicals

Hexamethylacetone, 98%, Thermo Scientific Chemicals

Catalog number: H53464.14
25 g, Each
Quantity
Catalog number: H53464.14
also known as H53464-14
Price (USD)
Price: 134.00
Online price: 113.65
Your price:
Quantity
-

Chemical Identifiers

CAS
815-24-7
IUPAC Name
2,2,4,4-tetramethylpentan-3-one
Molecular Formula
C9H18O
InChI Key
UIQGEWJEWJMQSL-UHFFFAOYSA-N
SMILES
CC(C)(C)C(=O)C(C)(C)C
Assay (HPLC)
97.5% min

Description

Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text