6-Chlor-1-Hexin, 98 %, Thermo Scientific Chemicals
6-Chlor-1-Hexin, 98 %, Thermo Scientific Chemicals
6-Chlor-1-Hexin, 98 %, Thermo Scientific Chemicals
6-Chlor-1-Hexin, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chlor-1-Hexin, 98 %, Thermo Scientific Chemicals

CAS: 10297-06-0 | C6H9Cl | 116.59 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 mL
25 mL
Katalognummer H53476.AC
auch als H53476-AC bezeichnet
Preis (EUR)
226,00
Each
Zum Warenkorb hinzufügen
Menge:
25 mL
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
226,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS10297-06-0
IUPAC Name6-chlorohex-1-yne
Molecular FormulaC6H9Cl
InChI KeyZUKOCGMVJUXIJA-UHFFFAOYSA-N
SMILESClCCCCC#C
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Infrared spectrumConforms to structure
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay from Supplier's CofA≥97.5%
6-Chloro-1-hexyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
6-Chlor-1-Hexyn wird als Zwischenprodukt der organischen chemischen Synthese verwendet.

Hinweise
Stabil unter empfohlenen Lagerbedingungen. Von Oxidationsmitteln entfernt aufbewahren
RUO – Research Use Only

Allgemeine Referenzen:

  1. Han-Cheng Zhang; Llorente V.R.Bonaga; Hong Ye; Claudia K.Derian; Bruce P.Damiano; Bruce E.Maryanoff. Novel bis(indolyl)maleimide pyridinophanes that are potent, selective inhibitors of glycogen synthase kinase-3. Bioorganic & Medicinal Chemistry Letters. 2007, 17, (10),2863-2868
  2. Edward Piers; J.Michael Chong. Reaction of Me3SnCu·Me2S with terminal alkynes. Preparation of 2-trimethylstannyl-1-alkenes. Canadian Journal of Chemistry. 1988, 66, (6),1425-1429