Methylmethansulfonat, 99%, Thermo Scientific Chemicals
Methylmethansulfonat, 99%, Thermo Scientific Chemicals
Methylmethansulfonat, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylmethansulfonat, 99%, Thermo Scientific Chemicals

CAS: 66-27-3 | C2H6O3S | 110.127 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer H55120.22
auch als H55120-22 bezeichnet
Preis (EUR)
615,00
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
615,00
Each
Chemikalien-Kennzeichnungen
CAS66-27-3
IUPAC Namemethyl methanesulfonate
Molecular FormulaC2H6O3S
InChI KeyMBABOKRGFJTBAE-UHFFFAOYSA-N
SMILESCOS(C)(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormLiquid
Assay (GC)>98.5%
Appearance (Color)Colorless to pale yellow
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
Methylmethansulfonat ist ein DNA-Addukt, das Dan unter 7-Guanin, 3-Guanin und 3-Adenin Methylgruppen hinzufügt.

Löslichkeit
Sehr gut in Wasser löslich.

Hinweise
Bei Raumtemperatur und entsprechender Belüftung aufbewahren. Von Hitze und Oxidationsmitteln fernhalten.
RUO – Research Use Only

Allgemeine Referenzen:

  1. D. Segerbäck.; C.J. Calleman.; L. Ehrenberg.; G. Löfroth.; S. Osterman-Golkar. Evaluation of genetic risks of alkylating agents IV. Quantitative determination of alkylated amino acids in haemoglobin as a measure of the dose after treatment of mice with methyl methanesulfonate. Mutation Research/Fundamental and Molecular Mechanisms of Mutagenesis. 1978, 49 (1),71-82.
  2. William F. Morgan.; James E. Cleaver. 3-Aminobenzamide synergistically increases sister-chromatid exchanges in cells exposed to methyl methanesulfonate but not to ultraviolet light .Mutation Research Letters. Mutation Research Letters, 104 (6),361-366.