Thermo Scientific Chemicals

6-Hydroxyindole, 98%, Thermo Scientific Chemicals

Catalog number: H55951.MD
250 mg, Each
Thermo Scientific Chemicals

6-Hydroxyindole, 98%, Thermo Scientific Chemicals

Catalog number: H55951.MD
250 mg, Each
Quantity
Catalog number: H55951.MD
also known as H55951-MD
Price (USD)
Price: 56.00
Online price: 47.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2380-86-1
IUPAC Name
1H-indol-6-ol
Molecular Formula
C8H7NO
InChI Key
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CNC2=C1
Assay (HPLC)
97.5% min

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text