Thermo Scientific Chemicals

Phenazine methosulfate, 98+%, Thermo Scientific Chemicals

Catalog number: H56718.14
25 g, Each
Thermo Scientific Chemicals

Phenazine methosulfate, 98+%, Thermo Scientific Chemicals

Catalog number: H56718.14
25 g, Each
Quantity
Catalog number: H56718.14
also known as H56718-14
Price (USD)
Price: 507.00
Online price: 442.65
Your price:
Quantity
-

Chemical Identifiers

CAS
299-11-6
IUPAC Name
5-methylphenazin-5-ium methyl sulfate
Molecular Formula
C14H14N2O4S
InChI Key
RXGJTUSBYWCRBK-UHFFFAOYSA-M
SMILES
COS([O-])(=O)=O.C[N+]1=C2C=CC=CC2=NC2=CC=CC=C12
Assay (HPLC)
98+%

Description

It is used with ascorbic acid to determine nitric oxide reductase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used with ascorbic acid to determine nitric oxide reductase activity.

Solubility
Soluble in water. (100g/L at20°C)

Notes
Light sensitive. Incompatible with oxidizing agents. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text