It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Es wird in der Medizin und als wichtiger Rohstoff und als Zwischenprodukt in der organischen Synthese, für Pharmazeutika, Agrochemikalien und Farbstoffe verwendet.
Löslichkeit
Löslich in Alkohol und Chloroform
Hinweise
Bei Raumtemperatur lagern. Von Oxidationsmitteln und Heizstoffen fernhalten.
RUO – Research Use Only
David L. Musso.; G. Faye Orr.; Felicia R. Cochran.; James L. Kelley.; Jeffrey L. Selph.; Greg C. Rigdon.; Barrett R. Cooper.; Michael L. Jones. Indanylidenes. 2. Design and Synthesis of (E)-2-(4-Chloro-6-fluoro-1-indanylidene)-N-methylacetamide, a Potent Antiinflammatory and Analgesic Agent without Centrally Acting Muscle Relaxant Activity. J. Med. Chem. 2003, 46 (3), 409-416.