Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H63411.22
100 g, Each
Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H63411.22
100 g, Each
Quantity
Catalog number: H63411.22
also known as H63411-22
Price (USD)
Price: 1,134.00
Online price: 981.65
Your price:
Quantity
-

Chemical Identifiers

CAS
84000-07-7
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)propanoic acid
Molecular Formula
C19H19NO4
InChI Key
JOFHWKQIQLPZTC-UHFFFAOYNA-N
SMILES
CC(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>94.0%

Description

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text