Nortropinon Hydrochlorid, 97 %, Thermo Scientific Chemicals
Nortropinon Hydrochlorid, 97 %, Thermo Scientific Chemicals
Nortropinon Hydrochlorid, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nortropinon Hydrochlorid, 97 %, Thermo Scientific Chemicals

CAS: 25602-68-0 | C7H12ClNO | 161.629 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer H64485.14
auch als H64485-14 bezeichnet
Preis (EUR)
106,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
106,00
Each
Chemikalien-Kennzeichnungen
CAS25602-68-0
IUPAC Name8-azabicyclo[3.2.1]octan-3-one hydrochloride
Molecular FormulaC7H12ClNO
InChI KeyMZQWQFWRSDNBPV-UHFFFAOYNA-N
SMILESCl.O=C1CC2CCC(C1)N2
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only