Thermo Scientific Chemicals

N-(Diphenylmethylene)glycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H66614.22
100 g, Each
Thermo Scientific Chemicals

N-(Diphenylmethylene)glycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H66614.22
100 g, Each
Quantity
Catalog number: H66614.22
also known as H66614-22
Price (USD)

Chemical Identifiers

CAS
81167-39-7
IUPAC Name
methyl 2-[(diphenylmethylidene)amino]acetate
Molecular Formula
C16H15NO2
InChI Key
PQTOLHHWLUCKSB-UHFFFAOYSA-N
SMILES
COC(=O)CN=C(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
≥97.5%
Melting Point
42-44?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text