Thermo Scientific Chemicals

Spironolactone, Thermo Scientific Chemicals

Catalog number: J60119.06
5 g, Each
Thermo Scientific Chemicals

Spironolactone, Thermo Scientific Chemicals

Catalog number: J60119.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60119.06
also known as J60119-06
Price (USD)
Price: 139.00
Online price: 119.65
Your price:
Quantity
-

Chemical Identifiers

CAS
52-01-7
IUPAC Name
(1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
Molecular Formula
C24H32O4S
InChI Key
LXMSZDCAJNLERA-ZHYRCANASA-N
SMILES
CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12
Form
White solid
Assay (unspecified)
≥ 97
Assay by NMR
Conform to structure
Loss on Drying
≤0.5%

Description

Spironolactone used as potassium sparing diuretic. Also used primarily to treat ascites in patients with liver disease, low-renin hypertension, hypokalemia, and Conn’s syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text