Ficin, fig tree latex
Thermo Scientific Chemicals

Ficin, fig tree latex

 
Have Questions?
KatalognummerMenge
J60136.14
auch als J60136-14 bezeichnet
25 g
Katalognummer J60136.14
auch als J60136-14 bezeichnet
Preis (EUR)
260,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
260,00
Each
Chemikalien-Kennzeichnungen
CAS9001-33-6
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)Titer: 400-1000 Milk clotting units/mg
Immobilized Ficin was specifically designed for cleavage of mouse IgG1 into F(ab')2 or Fab fragments. Ficin acts on bonds involving uncharged and/or aromatic amino acids. The immobilization of ficin enhances stability against denaturation, heat and autolysis. Immobilization also eliminates any potential for antibody-enzyme adducts that cause continued sample digestion. In addition to these advantages, immobilization allows the ficin to be reused.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only