Aceclofenac, 99%
Aceclofenac, 99%
Aceclofenac, 99%
Thermo Scientific Chemicals

Aceclofenac, 99%

CAS: 89796-99-6 | C16H13Cl2NO4 | 354.183 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
10 g
Katalognummer J60355.06
auch als J60355-06 bezeichnet
Preis (EUR)
67,83
Exklusiv online
79,80
Ersparnis 11,97 (15%)
Each
Zum Warenkorb hinzufügen
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
67,83
Exklusiv online
79,80
Ersparnis 11,97 (15%)
Each
Zum Warenkorb hinzufügen
Specifications
CAS89796-99-6
ChEBICHEBI:31159
Chemischer Name oder MaterialAceclofenac
FarbeWeiß
DOT-InformationenTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Mehr anzeigen
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.
RUO – Research Use Only