Amiodarone hydrochloride
Amiodarone hydrochloride
Amiodarone hydrochloride
Thermo Scientific Chemicals

Amiodarone hydrochloride

A non-selective ion channel blocker; also an antiarrhythmic agent | CAS: 19774-82-4 | C25H30ClI2NO3 | 681.778 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
25 g
Katalognummer J60456.03
auch als J60456-03 bezeichnet
Preis (EUR)
21,80
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
21,80
Each
Specifications
CAS19774-82-4
Chemischer Name oder MaterialAmiodarone hydrochloride
FarbeWeiß
EINECS-Nummer243-293-2
Formelmasse681.77
Mehr anzeigen
Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
RUO – Research Use Only

General References:

  1. Yong-Qiang Zhang; Rajini Rao. Global disruption of cell cycle progression and nutrient response by the antifungal agent amiodarone. Journal of Biological Chemistry. 2007, 282, (52), 37844-37853.
  2. Minhui Wang; Jiaojiao Shan; Qian Yang; Xianglei Ma; Sisi Jin; Xiaoke Guo; Qidong You; Yiqun Tang. Antiarrhythmic efficacy of CPUY102122, a multiple ion channel blocker, on rabbits with ischemia/reperfusion injury. Pharmacological reports : PR.2014, 66, (6), 1022-1030.