Thermo Scientific Chemicals

Fasudil dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: J60594.ME
500 mg, Each
Thermo Scientific Chemicals

Fasudil dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: J60594.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60594.ME
also known as J60594-ME
Price (USD)
Price: 376.00
Online price: 328.65
Your price:
Quantity
-

Chemical Identifiers

CAS
203911-27-7
IUPAC Name
5-(1,4-diazepane-1-sulfonyl)isoquinoline dihydrochloride
Molecular Formula
C14H19Cl2N3O2S
InChI Key
NOXXIYDYFSNHDF-UHFFFAOYSA-N
SMILES
Cl.Cl.O=S(=O)(N1CCCNCC1)C1=C2C=CN=CC2=CC=C1
Assay (HPLC)
>99.0%

Description

Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

Solubility
Soluble in water and dimethylsulfoxide.

Notes
Incompatible with strong oxidizing agents. Sore in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text