D-Glutamin, 99+ %
D-Glutamin, 99+ %
D-Glutamin, 99+ %
D-Glutamin, 99+ %
Thermo Scientific Chemicals

D-Glutamin, 99+ %

Kinase-Inhibitor; hat antivasospastische Eigenschaften
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
25 g
Katalognummer J60784.03
auch als J60784-03 bezeichnet
Preis (EUR)
35,36
Exklusiv online
41,60
Ersparnis 6,24 (15%)
Each
Zum Warenkorb hinzufügen
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
35,36
Exklusiv online
41,60
Ersparnis 6,24 (15%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS5959-95-5
IUPAC Name(2R)-2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-GSVOUGTGSA-N
SMILESN[C@H](CCC(N)=O)C(O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥99.0%
Kinase inhibitor; has antivasospastic properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only