(+/-)-Miconazole nitrate, 98+%
(+/-)-Miconazole nitrate, 98+%
(+/-)-Miconazole nitrate, 98+%
Thermo Scientific Chemicals

(+/-)-Miconazole nitrate, 98+%

Inhibits cytochrome P450-dependent 14a-demethylase | CAS: 22832-87-7 | C18H15Cl4N3O4 | 479.14 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
Katalognummer J60796.14
auch als J60796-14 bezeichnet
Preis (EUR)
387,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
387,00
Each
Specifications
CAS22832-87-7
Chemischer Name oder Material(+/-)-Miconazole nitrate
FarbeWeiß
EINECS-Nummer245-256-6
Formelmasse479.15
Mehr anzeigen
Inhibits cytochrome P450-dependent 14a-demethylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only