Thermo Scientific Chemicals

Dextran, MW ca 75,000, Thermo Scientific Chemicals

Catalog number: J60989.18
50 g, Each
Thermo Scientific Chemicals

Dextran, MW ca 75,000, Thermo Scientific Chemicals

Catalog number: J60989.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60989.18
also known as J60989-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
9004-54-0
IUPAC Name
Dextran
Molecular Formula
(C6H9O5)n(C6H10O5)m
SMILES
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
MDL Number
MFCD00130935
Form
White to off white powder
Ash
≤ 2%
Loss on Drying
≤ 7.0 %
pH
4.5 to 7. 0 (6% solution)
Solution Test
Cold water solubility (1%): Soluble

Description

Dextran, MW ca 75,000 is used as a processing aid, stabilizer, thickener, and surface finishing agent.. Further, it is used in soft center confections. In addition to this, it is used as a barley malt substitute and as a plasma volume expander.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides

Application

• Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)

• This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin

• Dextran can be used to reduce thrombosis in animal models

• It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules • Dextran can be used as an immobilization agent in biosensors

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text