Thermo Scientific Chemicals

Felodipine, Thermo Scientific Chemicals

Catalog number: J61195.MD
250 mg, Each
Thermo Scientific Chemicals

Felodipine, Thermo Scientific Chemicals

Catalog number: J61195.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61195.MD
also known as J61195-MD
Price (USD)
Quantity
-

Chemical Identifiers

CAS
72509-76-3
IUPAC Name
3-ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C18H19Cl2NO4
InChI Key
RZTAMFZIAATZDJ-UHFFFAOYNA-N
SMILES
CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(Cl)=C1Cl)C(=O)OC
Purity
≥95.0%
Mass Spec
Conforms
Form
Powder or crystals or crystalline powder
Appearance (Color)
Yellow to yellow-green

Description

Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

Solubility
Soluble in DMSO. Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text