Thermo Scientific Chemicals

Sodium pyruvate, Cell Culture Grade, Thermo Scientific Chemicals

Catalog number: J61840.22
100 g, Each
Thermo Scientific Chemicals

Sodium pyruvate, Cell Culture Grade, Thermo Scientific Chemicals

Catalog number: J61840.22
100 g, Each
Quantity
Catalog number: J61840.22
also known as J61840-22
Price (USD)
Price: 270.00
Online price: 229.65
Your price:
Quantity
-

Chemical Identifiers

CAS
113-24-6
IUPAC Name
sodium 2-oxopropanoate
Molecular Formula
C3H3NaO3
InChI Key
DAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES
[Na+].CC(=O)C([O-])=O
Assay (Titration)
≥99.0% (dried basis)
Loss on Drying
≤2.0%
Heavy Metals
≤10 ppm (as Pb)
pH
5.0-7.0
Appearance (Color)
White to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text