Azathioprine
Azathioprine
Azathioprine
Thermo Scientific Chemicals

Azathioprine

CAS: 446-86-6 | C9H7N7O2S | 277.262 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
10 g
Katalognummer J62314.09
auch als J62314-09 bezeichnet
Preis (EUR)
908,00
Each
Menge:
10 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
908,00
Each
Specifications
CAS446-86-6
ChEBICHEBI:2948
Chemischer Name oder MaterialAzathioprine
FarbeGelb
EINECS-Nummer207-175-4
Mehr anzeigen
Azathioprine is an immunosuppressive purine antimetabolite. The mechanism of action of azathioprine is as a nucleic acid synthesis inhibitor. It has shown promise in treatment of alopecia areata (autoimmune hair loss), with no difference in effectiveness between genders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Dichloromethane and dimethyl sulfoxide.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
RUO – Research Use Only
W.R. Connell.; Kamm.; Ritchie.; Prof J.E. Lennard-Jones.; Dickson.; A.M. Balkwill. Long-term neoplasia risk after azathioprine treatment in inflammatory bowel disease.The Lancet. 1994, 343 (8908),1249-1252 .