Thermo Scientific Chemicals

Flumazenil, 98%, Thermo Scientific Chemicals

Catalog number: J62537.ME
500 mg, Each
Thermo Scientific Chemicals

Flumazenil, 98%, Thermo Scientific Chemicals

Catalog number: J62537.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62537.ME
also known as J62537-ME
Price (USD)
Price: 1,213.00
Online price: 1,049.65
Your price:
Quantity
-

Chemical Identifiers

CAS
78755-81-4
IUPAC Name
ethyl 12-fluoro-8-methyl-9-oxo-2,4,8-triazatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaene-5-carboxylate
Molecular Formula
C15H14FN3O3
InChI Key
OFBIFZUFASYYRE-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=C2CN(C)C(=O)C3=CC(F)=CC=C3N2C=N1
Form
Powder
Assay from Suppliers CofA
≥97.5%

Description

Benzodiazepine antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text