Thermo Scientific Chemicals

Doxepin hydrochloride, Thermo Scientific Chemicals

Catalog number: J62579.14
25 g, Each
Thermo Scientific Chemicals

Doxepin hydrochloride, Thermo Scientific Chemicals

Catalog number: J62579.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62579.14
also known as J62579-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1229-29-4
IUPAC Name
hydrogen dimethyl({3-[(2E)-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene]propyl})amine chloride
Molecular Formula
C19H22ClNO
InChI Key
MHNSPTUQQIYJOT-SJDTYFKWSA-N
SMILES
[H+].[Cl-].CN(C)CC\C=C1/C2=CC=CC=C2COC2=CC=CC=C12
Assay (unspecified)
98.0% min.
Assay (unspecified)
Z-isomer: 13-18.5%
Loss on Drying
0.5% max.
Melting Point
185-191?C
Sulfated Ash
0.2% max.

Description

It is a highly potent H1 histamine receptor antagonist (Kd = 310 pM) and tricyclic antidepressant that inhibits the reuptake of serotonin and norepinephrine and acts as an antagonist at histamine, serotonin, adrenergic, and muscarinic receptors with Ki values in the nanomolar range. Also binds to the H4 histamine receptor (pKi = 6.79). Used clinically to treat anxiety and depression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text