Evodiamine
Evodiamine
Evodiamine
Evodiamine
Thermo Scientific Chemicals

Evodiamine

CAS: 518-17-2 | C19H17N3O, C19H17N3O | 303.37 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mg
250 mg
Katalognummer J62771.MC
auch als J62771-MC bezeichnet
Preis (EUR)
70,30
Each
Menge:
100 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
70,30
Each
Specifications
CAS518-17-2
BeschreibungOff-white to pale yellow
Zur Verwendung mit (Anwendung)A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
FormFest
Formelmasse303.40
Mehr anzeigen
A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Inhalt und Lagerung
-30°bis -10 °C