Tyrphostin A9, 99 %, Thermo Scientific Chemicals
Tyrphostin A9, 99 %, Thermo Scientific Chemicals
Tyrphostin A9, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tyrphostin A9, 99 %, Thermo Scientific Chemicals

Selektiver Inhibitor der PDGF-Rezeptor-Tyrosinkinase
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 mg
100 mg
Katalognummer J63058.MC
auch als J63058-MC bezeichnet
Preis (EUR)
211,00
Each
Menge:
100 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
211,00
Each
Specifications
CAS10537-47-0
ChEBICHEBI:82168
Chemischer Name oder MaterialTyrphostin A9
FarbeGelb
DOT-InformationenTransportgefahrenklasse:6.1; Verpackungsgruppe:III; richtiger Versandname:GIFTIGER FESTSTOFF, ORGANISCH, N.O.S.
Mehr anzeigen
Selective inhibitor of PDGF receptor tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only