Urapidil hydrochloride, 98+%
Urapidil hydrochloride, 98+%
Urapidil hydrochloride, 98+%
Thermo Scientific Chemicals

Urapidil hydrochloride, 98+%

alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist | CAS: 64887-14-5 | C20H29N5O3·HCl
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 mg
1 g
Katalognummer J63219.03
auch als J63219-03 bezeichnet
Preis (EUR)
191,00
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
191,00
Each
Specifications
Assay≥98%
CAS64887-14-5
BeschreibungWeiß
Zur Verwendung mit (Anwendung)alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist
FormPulver
Mehr anzeigen
alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only