(+)-cis-Diltiazem hydrochloride
(+)-cis-Diltiazem hydrochloride
(+)-cis-Diltiazem hydrochloride
Thermo Scientific Chemicals

(+)-cis-Diltiazem hydrochloride

An L-type calcium channel blocker | CAS: 33286-22-5 | C22H27ClN2O4S | 450.978 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
10 g
25 g
Katalognummer J63245.06
auch als J63245-06 bezeichnet
Preis (EUR)
135,00
Each
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
135,00
Each
Chemikalien-Kennzeichnungen
CAS33286-22-5
IUPAC Namehydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
Molecular FormulaC22H27ClN2O4S
InChI KeyHDRXZJPWHTXQRI-BHDTVMLSSA-N
SMILES[H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormPowder
Assay from Suppliers CofA≥94.0%
An L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only