Pektinase, Aspergillus niger
Pektinase, <i>Aspergillus niger</i>
Thermo Scientific Chemicals

Pektinase, Aspergillus niger

Wird in der Vorbereitung von Pflanzenprotoplast verwendet, um die Zellwand vor der Isolierung von Organellen zu verdauen. Pektinase erleichtert das Mazerieren von Pflanzenmaterialien und Extraktionsprozessen, einschließlich der Organellvorbereitung.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 mg
1 g
Katalognummer J63408.03
auch als J63408-03 bezeichnet
Preis (EUR)
450,00
Each
Zum Warenkorb hinzufügen
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
450,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS9032-75-1
SpecificationsSpecification SheetDatenblatt
Assay from Supplier's CofA≥20.0 units per mg dry weight (u/mg dw)
FormLyophilized powder
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only