L-Glutaminsäure-Mononatriumsalz
L-Glutaminsäure-Mononatriumsalz
L-Glutaminsäure-Mononatriumsalz
L-Glutaminsäure-Mononatriumsalz
Thermo Scientific Chemicals

L-Glutaminsäure-Mononatriumsalz

CAS: 142-47-2 | C5H8NNaO4 | 169.11 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
25 g
Katalognummer J63424.09
auch als J63424-09 bezeichnet
Preis (EUR)
45,00
Each
Zum Warenkorb hinzufügen
Menge:
10 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
45,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS142-47-2
IUPAC Namesodium 2-amino-4-carboxybutanoate
Molecular FormulaC5H8NNaO4
InChI KeyLPUQAYUQRXPFSQ-UHFFFAOYNA-M
SMILES[Na+].NC(CCC(O)=O)C([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormPowder or crystals or crystalline powder
Loss on Drying≤0.5%
Assay from Supplier's CofA≥98.0%
Appearance (Color)White to pale cream
Heavy Metals≤0.002%
L-Glutamic acid monosodium salt is used as a flavor enhancer, swine feed additive as well as to improve the taste of bitter drugs. It is used therapeutically to reduce blood ammonia levels. It serves as an excitatory amino acid neurotransmitter, which is an agonist at glur and NMDA receptors. Further, it is used to reduce the intake of sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Wasserlöslich. Unlöslich im Äther.

Hinweise
Nicht kompatibel mit starken Oxidationsmitteln.
RUO – Research Use Only

Allgemeine Referenzen:

  1. Jiang, L.; Pei, H.; Hu, W.; Ji, Y.; Han, L.; Ma, G. The feasibility of using complex wastewater from a monosodium glutamate factory to cultivate Spirulina subsalsa and accumulate biochemical composition. Bioresour. Technol. 2015, 180, 304-310.
  2. Matoušková, P.; Bártíková, H.; Boušová, I.; Levorová, L.; Szotáková, B.; Skálová, L. Drug-Metabolizing and Antioxidant Enzymes in Monosodium L-Glutamate Obese Mice. Drug Metab. Dispos. 2015, 43 (2), 258-265.