Bexarotene, 98%, Thermo Scientific Chemicals
Bexarotene, 98%, Thermo Scientific Chemicals
Bexarotene, 98%, Thermo Scientific Chemicals
Bexarotene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bexarotene, 98%, Thermo Scientific Chemicals

Synthetic retinoid analog with specific affinity for the retinoid X receptor
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mg
300 mg
Katalognummer J63701.MC
auch als J63701-MC bezeichnet
Preis (EUR)
130,00
Each
Zum Warenkorb hinzufügen
Menge:
100 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
130,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS153559-49-0
SpecificationsSpecification SheetDatenblatt
FormPowder or crystals or crystalline powder
Proton NMRConforms
Appearance (Color)White to pale yellow
Assay from Suppliers CofA≥97.5%
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer's model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer′s model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

Solubility
Soluble in DMSO and ethanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
Matthew N. Lowe.; Greg L. Plosker. Bexarotene. American Journal of Clinical Dermatology. 2000, 1, (4), 245-250.