Indapamide
Indapamide
Indapamide
Thermo Scientific Chemicals

Indapamide

CAS: 26807-65-8 | C16H16ClN3O3S | 365.83 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
Katalognummer J63846.03
auch als J63846-03 bezeichnet
Preis (EUR)
44,20
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
44,20
Each
Specifications
CAS26807-65-8
ChEBICHEBI:5893
Chemischer Name oder MaterialIndapamide
FarbeWeiß
EINECS-Nummer248-012-7
Mehr anzeigen
Used as an antihypertensive. Diuretic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used as an antihypertensive
  • Diuretic
RUO – Research Use Only