Amikacin disulfate
Amikacin disulfate
Amikacin disulfate
Thermo Scientific Chemicals

Amikacin disulfate

An aminoglycoside antibiotic derived from Kanamycin A | CAS: 39831-55-5 | C22H47N5O21S2 | 781.75 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer J63862.22
auch als J63862-22 bezeichnet
Preis (EUR)
445,00
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
445,00
Each
Chemikalien-Kennzeichnungen
CAS39831-55-5
SpecificationsSpecification SheetDatenblatt
pH2.0 to 4.0 (1% in water, 25?C)
Residue on ignition<1.0%
Assay (unspecified)674 to 786 ?g/mg
Loss on Drying<13%
Amikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.

Solubility
Soluble in water at 50mg/ml with warming

Notes
Potency: 674-786 micrograms per mg
RUO – Research Use Only