Thermo Scientific Chemicals

Phenol, ultrapure, 99%, unstab.

Catalog number: J64011.22
100 g, Each
Thermo Scientific Chemicals

Phenol, ultrapure, 99%, unstab.

Catalog number: J64011.22
100 g, Each
Quantity
Catalog number: J64011.22
also known as J64011-22
Price (USD)
Price: 44.40
Online price: 37.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-95-2
IUPAC Name
phenol
Molecular Formula
C6H6O
InChI Key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1
Residue on Evaporation
≤0.05%
Appearance (Color)
Colorless
Water Content (Karl Fischer Titration)
≤0.15%
Heavy Metals (as Pb)
≤5ppm
Form
Crystals or crystalline solid or fused solid (clear liquid as melt)

Description

Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.

Solubility
Soluble in water (82 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text