Taq DNA Polymerase Kit
Thermo Scientific Chemicals

Taq DNA Polymerase Kit

 
Have Questions?
KatalognummerMenge
J64594.XEX
auch als J64594-X! bezeichnet
400 units
Katalognummer J64594.XEX
auch als J64594-X! bezeichnet
Preis (EUR)
-
Menge:
400 units
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS9012-90-2
SpecificationsSpecification SheetDatenblatt
Assay from Supplier's CofANon-Specific DNase Activity: Pass
Assay from Supplier's CofAEndonuclease Activity: Pass
Assay from Supplier's CofAqPCR DNA Contamination: Pass
Catalyzes polymeration of deoxyribonucleotides into DNA strands. Suitable for PCR and automated sequencing reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only