Thermo Scientific Chemicals
Tripeptidyl Peptidase Inhibitor
Catalog number: J64760.MA
10 mg, Each
Thermo Scientific Chemicals
Tripeptidyl Peptidase Inhibitor
Catalog number: J64760.MA
10 mg, Each
Quantity
Catalog number: J64760.MA
also known as J64760-MA
Price (USD)
Price: 202.00
Online price: 176.65
Your price:
Quantity
-
Description
Tripeptidyl peptidase Inhibitor is a serine protease inhibitor which irreversibly inhibits TPPII (tripeptidyl peptidase II), a giant protease which may substitute for some proteasome functions, and reversibly inhibits Cln (TPPI). AAF-CMK shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II