PCR Mycoplasma detection kit
Thermo Scientific Chemicals

PCR Mycoplasma detection kit

The PCR Mycoplasma Detection Kit streamlines the process of identifying a mycoplasma outbreak in laboratory facilities.
Have Questions?
KatalognummerMenge
J66117.AMJ
auch als J66117-&J bezeichnet
100 μl
Katalognummer J66117.AMJ
auch als J66117-&J bezeichnet
Preis (EUR)
359,00
Each
Menge:
100 μl
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
359,00
Each
Specifications
Chemischer Name oder MaterialPCR Mycoplasma detection kit
Empfohlene LagerungLagerung bei -20 °C
LöslichkeitsinformationenSoluble in water.
Lagerungshinweis 1-30 °C bis -10 °C
TSCANo
Mehr anzeigen
Mycoplasma PCR Detection Kit has been used for screening of mycoplasma infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Mycoplasma are some of the smallest living organisms, making them difficult to identify

• Mycoplasma outbreaks are common and can devastate cell and tissue research overnight

• Alternative testing methods can be slow and inaccurate

Applications

• Highly specific for mycoplasma DNA—little to no cross-reactivity

• Suitable for identification of mycoplasma in cell and tissue cultures

RUO – Research Use Only

Literature references:

  1. Corral-Vázquez, C.; Aguilar-Quesada, R.; Catalina, P.; Lucena-Aguilar, G.; Ligero, G., Miranda, B.; Carrillo-Ávila, J.A. Cell lines authentication and mycoplasma detection as minimun quality control of cell lines in biobanking. Cell Tissue Bank. 2017,(2), 271-280. doi: 10.1007/s10561-017-9617-6. Epub 2017 Mar 2. PMID: 28255773 Drexler, H.G.; Uphoff, C.C. Mycoplasma contamination of cell cultures: incid

Häufig gestellte Fragen (FAQ)

What storage conditions should I use for the PCR Mycoplasma detection kit (Cat. No. J66117.AMJ)?

The PCR Mycoplasma detection kit should be stored at -20°C. We recommend that you avoid repeated freeze/thaw cycles of the master mix to retain maximum performance.