Tazobactam sodium
Tazobactam sodium
Tazobactam sodium
Thermo Scientific Chemicals

Tazobactam sodium

Beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases. | CAS: 89785-84-2 | C10H12N4NaO5S | 323.279 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mg
1 g
Katalognummer J66226.MC
auch als J66226-MC bezeichnet
Preis (EUR)
62,90
Each
Menge:
100 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
62,90
Each
Chemikalien-Kennzeichnungen
CAS89785-84-2
IUPAC Namesodium (2S,3S)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC10H11N4NaO5S
InChI KeyRFMIKMMOLPNEDG-LEFOPYBQSA-M
SMILES[Na+].C[C@]1(CN2C=CN=N2)[C@@H](N2C(CC2=O)S1(=O)=O)C([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormPowder or crystalline powder or crystals
Water Content (Karl Fischer Titration)≤2.5%
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98.0%
pH5.0-7.0
Tazobactam sodium is a beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only