Thermo Scientific Chemicals
Imipenem monohydrate, 95%, Thermo Scientific Chemicals
Catalog number: J66749.MB
25 mg, Each
Thermo Scientific Chemicals
Imipenem monohydrate, 95%, Thermo Scientific Chemicals
Catalog number: J66749.MB
25 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66749.MB
also known as J66749-MB
Price (USD)
Price: 247.00
Online price: 209.65
Your price:
Quantity
-
Description
Imipenem monohydrate is a broad spectrum B-lactam antibiotic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Imipenem monohydrate is a beta-lactam derived from the compound thienamycin, produced by Streptomyces cattleya
• Disrupts cell wall integrity, preventing cell division and promoting lysis by interfering with the activity of penicillin binding proteins
• Imipenem is a third-generation cephalosporin that can kill both gram-negative and gram-positive bacteria
Applications
• Suitable for the study of antibiotic resistance
• High-quality, laboratory grade for use in vitro experiments