Thermo Scientific Chemicals

Trimethoprim, 50 mg/ml in DMSO, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67175.8EQ
5 x 1 mL, Each
Thermo Scientific Chemicals

Trimethoprim, 50 mg/ml in DMSO, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67175.8EQ
5 x 1 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67175.8EQ
also known as J67175-8=
Price (USD)
Price: 504.00
Your price:
Quantity
-

Chemical Identifiers

CAS
738-70-5
IUPAC Name
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Formula
C14H18N4O3
InChI Key
IEDVJHCEMCRBQM-UHFFFAOYSA-N
SMILES
COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Transition Temperature
Trimethoprim: 50 mg/mL

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text