Thermo Scientific Chemicals

Nalidixic acid, 50 mg/ml in water, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67194.XF
1 mL, Each
Thermo Scientific Chemicals

Nalidixic acid, 50 mg/ml in water, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67194.XF
1 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67194.XF
also known as J67194-XF
Price (USD)
Price: 75.40
Online price: 65.65
Your price:
Quantity
-

Description

Nalidixic acid is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. It is used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependent)t DNA supercoiling, and chromosome fragmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalidixic acid is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. It is used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependent)t DNA supercoiling, and chromosome fragmentation.

Solubility
Soluble in chloroform (50 mg/ml), ethanol (0.9 mg/ml) at 23°C, methanol (1.3 mg/ml) at 23°C, toluene (1.6 mg/ml) at 23°C, DMSO (≥ 6 mg/ml), and solution of carbonates. Fully miscible in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text