Gold-Nanopartikel, 50 nm, geliefert in 0.1 mM PBS, reaktionsmittelfrei, 99 %, OD1, 535 nm-Absorption, Thermo Scientific Chemicals
Gold-Nanopartikel, 50 nm, geliefert in 0.1 mM PBS, reaktionsmittelfrei, 99 %, OD1, 535 nm-Absorption, Thermo Scientific Chemicals
Gold-Nanopartikel, 50 nm, geliefert in 0.1 mM PBS, reaktionsmittelfrei, 99 %, OD1, 535 nm-Absorption, Thermo Scientific Chemicals
Gold-Nanopartikel, 50 nm, geliefert in 0.1 mM PBS, reaktionsmittelfrei, 99 %, OD1, 535 nm-Absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold-Nanopartikel, 50 nm, geliefert in 0.1 mM PBS, reaktionsmittelfrei, 99 %, OD1, 535 nm-Absorption, Thermo Scientific Chemicals

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 mL
100 mL
Katalognummer J67432.AE
auch als J67432-AE bezeichnet
Preis (EUR)
-
Menge:
100 mL
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Buffer0.1 mM Phosphate-Buffered Saline (0.01X PBS)
CommentCore Size: 50nm (±3nm)
lambda-max (UV-Vis)531 - 535 nm
Particle Size~3.51E+10
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only