Thermo Scientific Chemicals

Dimethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: L00537.14
25 g, Each
Thermo Scientific Chemicals

Dimethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: L00537.14
25 g, Each
Quantity
Catalog number: L00537.14
also known as L00537-14
Price (USD)
Price: 390.00
Online price: 331.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4202-14-6
IUPAC Name
dimethyl (2-oxopropyl)phosphonate
Molecular Formula
C5H11O4P
InChI Key
UOWIYNWMROWVDG-UHFFFAOYSA-N
SMILES
COP(=O)(CC(C)=O)OC
Form
Liquid
Refractive Index
1.4365-1.4415 @ 20?C
Appearance (Color)
Clear colorless to white to yellow
Assay (GC)
≥96.0%

Description

Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text