Thermo Scientific Chemicals

Thiocarbohydrazide, 97%, Thermo Scientific Chemicals

Catalog number: L01205.22
100 g, Each
Thermo Scientific Chemicals

Thiocarbohydrazide, 97%, Thermo Scientific Chemicals

Catalog number: L01205.22
100 g, Each
Quantity
Catalog number: L01205.22
also known as L01205-22
Price (USD)
Price: 436.00
Online price: 370.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2231-57-4
IUPAC Name
1,3-diaminothiourea
Molecular Formula
CH6N4S
InChI Key
LJTFFORYSFGNCT-UHFFFAOYSA-N
SMILES
NNC(=S)NN
Assay (unspecified)
≥95.0%
Appearance (Color)
White to pale cream to pale brown or grey
Form
Crystals or powder or crystalline powder
Melting Point
159-180?C (dec)

Description

Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.

Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text