Thermo Scientific Chemicals

Dicyclopropylmethanol, 97%, Thermo Scientific Chemicals

Catalog number: L01816.03
1 g, Each
Thermo Scientific Chemicals

Dicyclopropylmethanol, 97%, Thermo Scientific Chemicals

Catalog number: L01816.03
1 g, Each
Quantity
Catalog number: L01816.03
also known as L01816-03
Price (USD)
Price: 40.90
Online price: 35.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14300-33-5
Refractive Index
1.4625-1.4675 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow

Description

Dicyclopropylmethanol is used as protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopropylmethanol is used as protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved.

Solubility
Sparingly soluble in water.

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text