Thermo Scientific Chemicals

Benzyl benzoate, 99+%, Thermo Scientific Chemicals

Catalog number: L03258.0B
1000 g, Each
Thermo Scientific Chemicals

Benzyl benzoate, 99+%, Thermo Scientific Chemicals

Catalog number: L03258.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03258.0B
also known as L03258-0B
Price (USD)
Price: 48.30
Online price: 41.65
Your price:
Quantity
-

Chemical Identifiers

CAS
120-51-4
IUPAC Name
benzyl benzoate
Molecular Formula
C14H12O2
InChI Key
SESFRYSPDFLNCH-UHFFFAOYSA-N
SMILES
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to white to yellow
Identification (FTIR)
Conforms
Refractive Index
1.5680-1.5700 @ 20?C
Form
Liquid
Assay (GC)
≥99.0%

Description

Benzyl benzoate is used as repellent for chiggers, ticks, and mosquitoes. It is an effective and inexpensive topical treatment for human scabies. It has vasodilating and spasmolytic effects and is present in many asthma and whooping cough drugs. It is also a dye carrier, solvent for cellulose derivatives, plasticizer, and fixative in the perfume industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with ethanol, alcohol, chloroform, ether, oils.

Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text