Thermo Scientific Chemicals

Isobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: L04038.AP
500 mL, Each
Thermo Scientific Chemicals

Isobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: L04038.AP
500 mL, Each
Quantity
Catalog number: L04038.AP
also known as L04038-AP
Price (USD)
Price: 38.60
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
79-31-2
IUPAC Name
2-methylpropanoic acid
Molecular Formula
C4H8O2
InChI Key
KQNPFQTWMSNSAP-UHFFFAOYSA-N
SMILES
CC(C)C(O)=O
Assay (GC)
≥98.5% (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.3915-1.3950 @ 20°C (non-U.S. specification)
Assay from Suppliers CofA
≥98.5% (U.S. specification)
Form
Liquid

Description

Isobutyric acid is used to prepare esters for flavors and perfumes. It is also used as a disinfecting agent, preservative and tanning agent. It finds applications in textile, varnish and the leather industry. Further, it is used as a lactation stimulant in dairy cattle. In addition to this, it is used in the preparation of tetramethylsuccinic acid and diisopropyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyric acid is used to prepare esters for flavors and perfumes. It is also used as a disinfecting agent, preservative and tanning agent. It finds applications in textile, varnish and the leather industry. Further, it is used as a lactation stimulant in dairy cattle. In addition to this, it is used in the preparation of tetramethylsuccinic acid and diisopropyl ketone.

Solubility
Miscible with water, alcohol, chloroform and ether. Slightly miscible with carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text