Thermo Scientific Chemicals

Polygalacturonic acid, M.W. 25,000-50,000, Thermo Scientific Chemicals

Catalog number: L04206.36
500 g, Each
Thermo Scientific Chemicals

Polygalacturonic acid, M.W. 25,000-50,000, Thermo Scientific Chemicals

Catalog number: L04206.36
500 g, Each
Quantity
Catalog number: L04206.36
also known as L04206-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
25990-10-7
Molecular Formula
(C6H8O6)n
SMILES
O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O
MDL Number
MFCD00131972
Form
Powder
Identification (FTIR)
Conforms
pH
2.5-3.5 (1% suspension)
Appearance (Color)
White to cream
Assay (unspecified)
≥86.0% (dry wt. basis)

Description

Polygalacturonic acid is reagent which sequesters metal ions and produces gels that can be used for the production of biomolecules. It is used for drug release or to immobilize cells and enzymes . Polygalacturonic acid may be used to identify, differentiate and characterized plant cell wall degrading polygalacturonase(s) from fungi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text