Thermo Scientific Chemicals

n-Triacontane, 98+%, Thermo Scientific Chemicals

Catalog number: L05386.03
1 g, Each
Thermo Scientific Chemicals

n-Triacontane, 98+%, Thermo Scientific Chemicals

Catalog number: L05386.03
1 g, Each
Quantity
Catalog number: L05386.03
also known as L05386-03
Price (USD)
Price: 61.50
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
638-68-6
IUPAC Name
triacontane
Molecular Formula
C30H62
InChI Key
JXTPJDDICSTXJX-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Appearance (Color)
White
Form
Crystalline or waxy flakes or fused solid
Assay (GC)
≥98.0%
Melting Point (clear melt)
63.5-69.5?C
Identification (FTIR)
Conforms

Description

n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.

Solubility
Soluble in benzene, chloroform and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text