Thermo Scientific Chemicals

Methyl 2,6-difluorobenzoate, 97%, Thermo Scientific Chemicals

Catalog number: L08063.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2,6-difluorobenzoate, 97%, Thermo Scientific Chemicals

Catalog number: L08063.06
5 g, Each
Quantity
Catalog number: L08063.06
also known as L08063-06
Price (USD)
Price: 38.90
Online price: 33.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13671-00-6
IUPAC Name
methyl 2,6-difluorobenzoate
Molecular Formula
C8H6F2O2
InChI Key
QNPFLTKQLFSKBY-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(F)C=CC=C1F
Form
Liquid
Refractive Index
1.4730-1.4790 @ 20?C
Appearance (Color)
Clear colorless to white to yellow to pale brown
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text