Thermo Scientific Chemicals

3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals

Catalog number: L08349.03
1 g, Each
Thermo Scientific Chemicals

3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals

Catalog number: L08349.03
1 g, Each
Quantity
Catalog number: L08349.03
also known as L08349-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
17260-71-8
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
145.0-151.0?C

Description

It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text